missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 Spike RBD Antibody (B001Y) - (RBD) - BSA Free, Novus Biologicals™

Human Monoclonal Antibody

Brand:  Novus Biologicals NBP3-05725

Product Code. 18622907

  • £420.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SARS-CoV-2 Spike RBD Monoclonal antibody specifically detects SARS-CoV-2 Spike RBD in SARS-CoV-2 samples. It is validated for Western Blot, ELISA, Neutralization
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SARS-CoV-2 Spike RBD
Monoclonal
Unconjugated
PBS, 10% glycerol, pH7.2
Human
Protein A purified
RUO
Primary
SARS-CoV-2
Purified
Western Blot, ELISA, Neutralization
B001Y
Western Blot 1:1000-2000, ELISA 1:320000, Block/Neutralize
2019-nCoV Peplomer protein, 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, S protein, SARS-CoV-2, SARS-COV-2 E2, SARS-COV-2 Peplomer protein, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike, Spike glycoprotein, Spike S1, Spike S1 RBD, surface glycoprotein
SARS-CoV-2 Spike RBD recombinant protein
50 μg
Infections (Virus Bacteria and Parasites)
43740568
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.