missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 Spike RBD Antibody (2355) - Azide and BSA Free, Novus Biologicals™

Product Code. 18399014 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399014 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18399014 Supplier Novus Biologicals Supplier No. NBP307067

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

SARS-CoV-2 Spike RBD Monoclonal specifically detects SARS-CoV-2 Spike RBD in SARS-CoV-2 samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 Spike RBD
Applications ELISA
Classification Monoclonal
Clone 2355
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA
Formulation 0.01 M PBS (pH 7.2 - 7.4), 150 mM NaCl
Gene Alias 2019-nCoV Peplomer protein, 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, S protein, SARS-CoV-2, SARS-COV-2 E2, SARS-COV-2 Peplomer protein, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike, Spike glycoprotein, Spike S1, Spike S1 RBD, surface glycoprotein
Host Species Human
Immunogen Sequenced from human survivors of COVID-19 (SARS-CoV-2). The spike RBD is expressed on the surface of SARS-CoV-2.
Purification Method >95% pure by SDS-PAGE
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740568
Target Species SARS-CoV-2
Content And Storage Store at 4°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.