missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS-CoV-2 Spike Protein S2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114534
This item is not returnable.
View return policy
Description
SARS-CoV-2 Spike Protein S2 Polyclonal Antibody for Western Blot, ELISA
SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. SARS-CoV-2 Spike Protein is composed of S1 domain and S2 domain (Spike Protein). The S2 domain plays a role in fusion. Further, S proteins plays an important role in the induction of neutralizing-antibodies and T-cell responses, as well as protective immunity. CoV diversity is reflected in the variable Spike proteins (S proteins), which have evolved into forms differing in their receptor interactions and their response to various environmental triggers of virus-cell membrane fusion.
Specifications
| SARS-CoV-2 Spike Protein S2 | |
| Polyclonal | |
| Unconjugated | |
| Coronavirus; COVID; Covid 19; COVID19; COVID-19; COVID-19 S2; COVID19 Spike S2; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 | |
| Anti-SARS-CoV-2 (COVID-19) Spike antibody was raised against a peptide corresponding to 16 amino acids near the center terminus of SARS-CoV-2 (COVID-19) Spike glycoprotein. The immunogen is located within 900-950 amino acids of SARS-CoV-2 (COVID-19) Spike protein. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction