missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS-CoV-2 Spike (N501Y Mutant) Recombinant Rabbit Monoclonal Antibody (HL1425)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547040
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
The spike protein in SARS-CoV-2 (SARS-2-S) interacts with the human ACE2 receptor to gain entry into a cell to initiate infection SARS-2-S is cleaved into S1 and S2 subunits, with S1 serving the function of receptor-binding and S2 serving the function of membrane fusion The S protein undergoes two crucial cleavage events, with the first splitting S1 and S2 and the second splitting S2 into FP and S2' [PMID: 33466921].
Specifications
| SARS-CoV-2 Spike (N501Y Mutant) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| Western Blot | |
| HL1425 | |
| Unconjugated | |
| 2019-nCoV Spike; COVID-19 Spike; SARS-CoV-2 Spike | |
| Carrier-protein conjugated synthetic peptide surrounding mutant N501Y of SARS-CoV-2 (COVID-19) Spike. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction