missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Spike Llama anti-SARS-CoV, SARS-CoV-2, Clone: T3P2D11, Novus Biologicals™
Llama Monoclonal Antibody
£176.00 - £610.00
Specifications
| Antigen | SARS-CoV-2 Spike |
|---|---|
| Clone | T3P2D11 |
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18351402
|
Bio-Techne
NBP3-18524-0.025MG |
0.025 mg |
£176.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18392807
|
Novus Biologicals
NBP3-18524-0.1MG |
0.1 mg |
£610.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SARS-CoV-2 Spike Monoclonal antibody specifically detects SARS-CoV-2 Spike in SARS-CoV, SARS-CoV-2 samples. It is validated for ELISASpecifications
| SARS-CoV-2 Spike | |
| ELISA | |
| Unconjugated | |
| Llama | |
| Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| PBS | |
| 43740568 | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| T3P2D11 | |
| Monoclonal | |
| Purified | |
| RUO | |
| SARS-CoV, SARS-CoV-2 | |
| 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein, surface glycoprotein | |
| SARS-CoV-2 variant Spike proteins | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title