missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 Spike Antibody, HRP, Novus Biologicals™

Product Code. 18377454 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377454 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377454 Supplier Novus Biologicals Supplier No. NBP312853H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 Spike Polyclonal antibody specifically detects SARS-CoV-2 Spike in SARS-CoV-2 samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 Spike
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate HRP
Formulation PBS without Preservative
Gene Alias 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein, surface glycoprotein
Host Species Rabbit
Immunogen Anti-SARS-CoV-2 Spike 156-157EFdel (Delta Variant) antibody was raised against a peptide corresponding to 11 amino acids near the amino terminal domain of SARS-CoV-2 Delta variant (B.1.617.2) Spike protein. The immunogen is located within 130-180 amino acids of SARS-CoV-2 Delta variant (B.1.617.2) Spike protein.
Purification Method Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 43740568
Target Species SARS-CoV-2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.