missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 Spike Antibody (7A4D12), DyLight 488, Novus Biologicals™

Product Code. 18365293
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18365293 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18365293 Supplier Bio-Techne Supplier No. NBP312854G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SARS-CoV-2 Spike Monoclonal antibody specifically detects SARS-CoV-2 Spike in SARS-CoV-2 samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 Spike
Applications Western Blot, ELISA
Classification Monoclonal
Clone 7A4D12
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein, surface glycoprotein
Host Species Mouse
Immunogen Anti-SARS-CoV-2 (COVID-19) P681H Mutant Specific Spike antibody was raised against a peptide corresponding to 12 amino acids near the center of SARS-CoV-2 Alpha variant (B.1.1.7) Spike protein. The immunogen is located within 650-700 amino acids of SARS-CoV-2 Alpha Variant (B.1.1.7, UK) Spike protein.
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 43740568
Target Species SARS-CoV-2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.