missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 S protein (RBD, 319-541 aa) Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | SARS-CoV-2 S protein (RBD, 319-541 aa) |
|---|---|
| Clone | 1H3E9 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236392
|
Proteintech
67758-1-IG-20UL |
20 μL |
£135.00 £119.00 / 20µL Save £16.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30235957
|
Proteintech
67758-1-IG-150UL |
150 μL |
£349.00 £295.00 / 150µL Save £54.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SARS-CoV-2 S protein (RBD, 319-541 aa) antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of virus SARS-CoV-2 S protein (RBD, 319-541 aa). This antibody recognizes virus antigen. The SARS-CoV-2 S protein (RBD, 319-541 aa) antibody has been validated for the following applications: WB, ELISA, Neutralization analysis.Specifications
| SARS-CoV-2 S protein (RBD, 319-541 aa) | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 43740568 | |
| IgG1 | |
| Protein A purification | |
| Monoclonal Antibody |
| 1H3E9 | |
| Western Blot, ELISA, Neutralization | |
| Unconjugated | |
| Mouse | |
| Virus | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title