missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 ORF9c Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18397273 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397273 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18397273 Supplier Novus Biologicals Supplier No. NBP311930AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 ORF9c Polyclonal antibody specifically detects SARS-CoV-2 ORF9c in SARS-CoV-2 samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 ORF9c
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 2019-nCoV ORF9c, COVID-19 ORF9c, ORF14, ORF9c, ORF9c protein, Uncharacterized protein 14
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 16 amino acids near the amino-terminus of SARS-CoV-2 ORF9c protein. The immunogen is located within the first 50 amino acids of the SARS-CoV-2 ORF9c protein.
Purification Method Peptide Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Target Species SARS-CoV-2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.