missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SARS-CoV-2 ORF9b Polyclonal antibody specifically detects SARS-CoV-2 ORF9b in SARS-CoV-2 samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | SARS-CoV-2 ORF9b |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | HRP |
| Formulation | PBS without Preservative |
| Gene Alias | Accessory protein 9b, ORF9b, ORF-9b, Protein 9b |
| Host Species | Rabbit |
| Immunogen | This antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of SARS-CoV-2 (COVID-19) ORF9b protein. The immunogen is located in the first 50 amino acids of the SARS-CoV-2 (COVID-19) ORF9b protein. |
| Purification Method | Peptide Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?