missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 ORF9b Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18347363
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347363 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347363 Supplier Bio-Techne Supplier No. NBP307036AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 ORF9b Polyclonal antibody specifically detects SARS-CoV-2 ORF9b in SARS-CoV-2 samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 ORF9b
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Accessory protein 9b, ORF9b, ORF-9b, Protein 9b
Host Species Rabbit
Immunogen This antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of SARS-CoV-2 (COVID-19) ORF9b protein. The immunogen is located in the first 50 amino acids of the SARS-CoV-2 (COVID-19) ORF9b protein.
Purification Method Peptide Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Target Species SARS-CoV-2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.