missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 ORF7a Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18366722 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18366722 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18366722 Supplier Novus Biologicals Supplier No. NBP311936AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 ORF7a Polyclonal antibody specifically detects SARS-CoV-2 ORF7a in SARS-CoV-2 samples. It is validated for ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 ORF7a
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 2019-nCoV ORF7a, 2019-nCoV ORF7a Protein, Accessory Protein 7a, COVID-19 Accessory Protein 7a, COVID-19 ORF7a, COVID-19 Protein U122, COVID-19 Protein X4, Human coronavirus ORF7a Protein, ORF7a Protein, Protein U122, Protein X4, SARS-CoV-2 Accessory Protein 7a, SARS-CoV-2 Protein U122, SARS-CoV-2 Protein X4
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 9 amino acids near the carboxyl terminus of SARS-CoV-2 ORF7a protein. The immunogen is located within the last 50 amino acids of the SARS-CoV-2 ORF7a protein.
Purification Method Peptide Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 43740573
Target Species SARS-CoV-2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.