missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 ORF3b Antibody, DyLight 755, Novus Biologicals™

Product Code. 18331172 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18331172 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18331172 Supplier Novus Biologicals Supplier No. NBP311928IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 ORF3b Polyclonal antibody specifically detects SARS-CoV-2 ORF3b in SARS-CoV-2 samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 ORF3b
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 2019-nCoV ORF3b, Acessary protein 3b, COVID-19 ORF3b, Non-structural protein 3b, ns3b, Orf3b, ORF3b protein, Protein X2
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 16 amino acids near the central domain of SARS-CoV-2 ORF3b protein. The immunogen is located within 30-80 amino acids of the SARS-CoV-2 ORF3b protein.
Purification Method Peptide Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Target Species SARS-CoV-2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.