missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 ORF10 Antibody, DyLight 650, Novus Biologicals™

Product Code. 18375193 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18375193 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18375193 Supplier Novus Biologicals Supplier No. NBP311939C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 ORF10 Polyclonal antibody specifically detects SARS-CoV-2 ORF10 in SARS-CoV-2 samples. It is validated for ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 ORF10
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 2019-nCoV ORF10, 2019-nCoV ORF10 Protein, COVID-19 ORF10, Human coronavirus ORF10 Protein, ORF10, ORF10 protein, SARS-CoV-2, SARSCoV2 ORF10 Protein, SARS-CoV-2 ORF10 Protein, Severe Acute Respiratory Syndrome Coronavirus 2 ORF10 Protein
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 13 amino acids near the carboxyl terminus of SARS-CoV-2 ORF10 protein. The immunogen is located within the last 50 amino acids of the SARS-CoV-2 ORF10 protein.
Purification Method Peptide Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 43740576
Target Species SARS-CoV-2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.