missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SARS-CoV-2 ORF10 Polyclonal antibody specifically detects SARS-CoV-2 ORF10 in SARS-CoV-2 samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | SARS-CoV-2 ORF10 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1000, ELISA |
| Formulation | PBS, 10% glycerol, pH7.2 |
| Gene Alias | 2019-nCoV ORF10, 2019-nCoV ORF10 Protein, COVID-19 ORF10, Human coronavirus ORF10 Protein, ORF10, ORF10 protein, SARS-CoV-2, SARSCoV2 ORF10 Protein, SARS-CoV-2 ORF10 Protein, Severe Acute Respiratory Syndrome Coronavirus 2 ORF10 Protein |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide |
| Purification Method | Epitope affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?