missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Omicron (XBB.1) Spike Antibody (D008) [Janelia Fluor™ 585], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18853JF585
This item is not returnable.
View return policy
Description
SARS-CoV-2 Omicron (XBB.1) Spike Monoclonal antibody specifically detects SARS-CoV-2 Omicron (XBB.1) Spike in SARS-CoV-2 samples. It is validated for ELISA
Specifications
| SARS-CoV-2 Omicron (XBB.1) Spike | |
| Monoclonal | |
| Janelia Fluor 585 | |
| Human | |
| Protein A purified | |
| RUO | |
| SARS-CoV-2 | |
| Purified |
| ELISA | |
| D008 | |
| 50mM Sodium Borate | |
| This is a chimeric monoclonal antibody combining the constant domains of the human IgG1 molecule with mouse variable regions. The variable region was obtained from a mouse immunized with purified, recombinant SARS-CoV-2 Omicron XBB.1 Spike ECD Protein. The antibody was produced using recombinant antibody technology. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction