missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 Nucleocapsid Mouse anti-SARS-CoV, SARS-CoV-2, Clone: B3449M, Novus Biologicals™

Product Code. 18356886 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
200 μL
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18356886 200 μL 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18356886 Supplier Novus Biologicals Supplier No. NBP300514200UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SARS-CoV-2 Nucleocapsid Monoclonal specifically detects SARS-CoV-2 Nucleocapsid in SARS-CoV, SARS-CoV-2 samples. It is validated for Western Blot, ELISA, Lateral Flow Assay, ELISA Detection (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 Nucleocapsid
Applications Western Blot, ELISA, Lateral Flow
Classification Monoclonal
Clone B3449M
Concentration 4.5 mg/mL
Conjugate Unconjugated
Formulation Phosphate Buffered Saline, pH 7.4
Gene Alias 2019-nCoV N Protein, 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, COVID-19 nucleocapsid protein, Human coronavirus Nucleoprotein, Nucleocapsid protein, Nucleoprotein, SARS-CoV-2 Nucleocapsid protein, SARS-CoV-2 N protein, SARSCoV2 Nucleoprotein, Severe Acute Respiratory Syndrome Coronavirus 2 Nucleocapsid Protein
Host Species Mouse
Immunogen Recombinant SARS-CoV-2 Nucleocapsid Protein.
Purification Method Protein G purified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species SARS-CoV, SARS-CoV-2
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.