missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 Nucleocapsid Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18359672 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18359672 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18359672 Supplier Novus Biologicals Supplier No. NBP300510AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 Nucleocapsid Polyclonal antibody specifically detects SARS-CoV-2 Nucleocapsid in SARS-CoV-2, Virus samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 Nucleocapsid
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 2019-nCoV N Protein, 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, COVID-19 nucleocapsid protein, Human coronavirus Nucleoprotein, nucleocapsid phosphoprotein, Nucleocapsid protein, Nucleoprotein, ORF9; structural protein, SARS-CoV-2, SARS-CoV-2 Nucleocapsid protein, SARS-CoV-2 N protein, SARSCoV2 Nucleoprotein, Severe Acute Respiratory Syndrome Coronavirus 2 Nucleocapsid Protein
Host Species Rabbit
Immunogen Full length recombinant SARS-CoV-2 nucleocapsid protein.
Purification Method Immunogen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 43740575
Target Species SARS-CoV-2, Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.