missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 Nucleocapsid Antibody (4Z6T2), HRP, Novus Biologicals™

Product Code. 18384415 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384415 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18384415 Supplier Novus Biologicals Supplier No. NBP305765H

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 Nucleocapsid Polyclonal antibody specifically detects SARS-CoV-2 Nucleocapsid in SARS-CoV-2 samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 Nucleocapsid
Applications ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate HRP
Dilution ELISA
Formulation PBS
Gene Alias 2019-nCoV N Protein, 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, COVID-19 nucleocapsid protein, Human coronavirus Nucleoprotein, nucleocapsid phosphoprotein, Nucleocapsid protein, Nucleoprotein, ORF9; structural protein, SARS-CoV-2, SARS-CoV-2 Nucleocapsid protein, SARS-CoV-2 N protein, SARSCoV2 Nucleoprotein, Severe Acute Respiratory Syndrome Coronavirus 2 Nucleocapsid Protein
Host Species Rabbit
Immunogen E.coli derived SARS-COV-2 Nucleoproteins Ser2-Ala419(Gly335Ala) Accession #QHD43423.2
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 43740575
Target Species SARS-CoV-2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.