missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Nucleocapsid Antibody (4D0J7), mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
SARS-CoV-2 Nucleocapsid Monoclonal antibody specifically detects SARS-CoV-2 Nucleocapsid in SARS-CoV-2 samples. It is validated for ELISA, ELISA (Matched Antibody Pair)
Specifications
Specifications
| Conjugate | mFluor Violet 450 SE |
| Gene Alias | 2019-nCoV N Protein, 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, COVID-19 nucleocapsid protein, Human coronavirus Nucleoprotein, nucleocapsid phosphoprotein, Nucleocapsid protein, Nucleoprotein, ORF9; structural protein, SARS-CoV-2, SARS-CoV-2 Nucleocapsid protein, SARS-CoV-2 N protein, SARSCoV2 Nucleoprotein, Severe Acute Respiratory Syndrome Coronavirus 2 Nucleocapsid Protein |
| Immunogen | HK293 derived SARS-COV-2 Nucleocapsid Ser2-Ala419 Accession #YP_009724397.2 |
| Quantity | 0.1 mL |
| Content And Storage | Store at 4C in the dark. |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?