missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SARS-CoV-2 Nucleocapsid Monoclonal specifically detects SARS-CoV-2 Nucleocapsid in SARS-CoV-2 samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Sandwich ELISA.
Specifications
Specifications
| Antigen | SARS-CoV-2 Nucleocapsid |
| Applications | ELISA, Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Clone | 3868 |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:20-1:200, Immunocytochemistry/Immunofluorescence, Sandwich ELISA |
| Formulation | PBS (0.01M, pH 7.2) |
| Gene Alias | 2019-nCoV N Protein, 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, COVID-19 nucleocapsid protein, Human coronavirus Nucleoprotein, nucleocapsid phosphoprotein, Nucleocapsid protein, Nucleoprotein, ORF9; structural protein, SARS-CoV-2, SARS-CoV-2 Nucleocapsid protein, SARS-CoV-2 N protein, SARSCoV2 Nucleoprotein, Severe Acute Respiratory Syndrome Coronavirus 2 Nucleocapsid Protein |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a recombinant, full-length nucleoprotein. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?