missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 Nucleocapsid Antibody (040), Novus Biologicals™

Rabbit Monoclonal Antibody

£349.00 - £523.00

Specifications

Antigen SARS-CoV-2 Nucleocapsid
Clone 040
Dilution Western Blot 1:1000-1:10000, ELISA 1:5000-1:10000, Immunocytochemistry/ Immunofluorescence 1:20-1:100
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18631689
View Documents
Novus Biologicals
NBP3-12736-50ul
50 μg
£349.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18614896
View Documents
Novus Biologicals
NBP3-12736-100ul
100 μg
£523.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SARS-CoV-2 Nucleocapsid Monoclonal antibody specifically detects SARS-CoV-2 Nucleocapsid in SARS-CoV samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

SARS-CoV-2 Nucleocapsid
Western Blot 1:1000-1:10000, ELISA 1:5000-1:10000, Immunocytochemistry/ Immunofluorescence 1:20-1:100
Monoclonal
Purified
RUO
SARS-CoV
2019-nCoV N Protein, 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, COVID-19 nucleocapsid protein, Human coronavirus Nucleoprotein, nucleocapsid phosphoprotein, Nucleocapsid protein, Nucleoprotein, ORF9; structural protein, SARS-CoV-2, SARS-CoV-2 Nucleocapsid protein, SARS-CoV-2 N protein, SARSCoV2 Nucleoprotein, Severe Acute Respiratory Syndrome Coronavirus 2 Nucleocapsid Protein
Recombinant SARS-CoV-2 Nucleocapsid Protein
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
040
Western Blot, ELISA, Immunofluorescence
Unconjugated
Rabbit
Virology Bacteria and Parasites
0.2 um filtered solution in PBS
43740575
IgG
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.