missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SARS-CoV-2 nsp9 Polyclonal antibody specifically detects SARS-CoV-2 nsp9 in SARS-CoV-2 samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence
Specifications
Specifications
| Antigen | SARS-CoV-2 nsp9 |
| Applications | ELISA, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 755 |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | Non-structural protein 9, nsp9 replicase |
| Host Species | Rabbit |
| Immunogen | This antibody was raised against a peptide corresponding to 13 amino acids near the center of SARS-CoV-2 (COVID-19) NSP9 protein. The immunogen is located within 30-80 amino acids of the SARS-CoV-2 (COVID-19) NSP9 protein. |
| Purification Method | Peptide Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?