missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 nsp7 Antibody, FITC, Novus Biologicals™
Description
SARS-CoV-2 nsp7 Polyclonal antibody specifically detects SARS-CoV-2 nsp7 in SARS-CoV-2 samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence
Specifications
Specifications
| Antigen | SARS-CoV-2 nsp7 |
| Applications | ELISA, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | FITC |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | Non-structural protein 7, nsp7 replicase |
| Host Species | Rabbit |
| Immunogen | This antibody was raised against a peptide corresponding to 12 amino acids near the carboxyl terminus of the SARS-CoV-2 (COVID-19) NSP7 protein. The immunogen is located within the last 50 amino acids of the SARS-CoV-2 (COVID-19) NSP7 protein. |
| Purification Method | Peptide Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?