missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 nsp7 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18342363 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342363 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18342363 Supplier Novus Biologicals Supplier No. NBP307048AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 nsp7 Polyclonal antibody specifically detects SARS-CoV-2 nsp7 in SARS-CoV-2 samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 nsp7
Applications ELISA, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Non-structural protein 7, nsp7 replicase
Host Species Rabbit
Immunogen This antibody was raised against a peptide corresponding to 12 amino acids near the carboxyl terminus of the SARS-CoV-2 (COVID-19) NSP7 protein. The immunogen is located within the last 50 amino acids of the SARS-CoV-2 (COVID-19) NSP7 protein.
Purification Method Peptide Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 43740578
Target Species SARS-CoV-2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.