missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 nsp6 Antibody, DyLight 650, Novus Biologicals™

Product Code. 18324862 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18324862 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18324862 Supplier Novus Biologicals Supplier No. NBP307049C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 nsp6 Polyclonal antibody specifically detects SARS-CoV-2 nsp6 in SARS-CoV-2 samples. It is validated for ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 nsp6
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Non-structural protein 6
Host Species Rabbit
Immunogen This antibody was raised against a peptide corresponding to 18 amino acids in the center of SARS-CoV-2 (COVID-19) NSP6 protein. The immunogen is located in between 230-280 amino acids of the SARS-CoV-2 (COVID-19) NSP6 protein.
Purification Method Peptide Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 43740578
Target Species SARS-CoV-2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.