missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 nsp3 Rabbit anti-SARS-CoV-2, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £299.00
Specifications
| Antigen | SARS-CoV-2 nsp3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18306875
|
Novus Biologicals
NBP3-15986-20UL |
20 μg |
£139.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18331367
|
Novus Biologicals
NBP3-15986-100UL |
100 μg |
£299.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SARS-CoV-2 nsp3 Polyclonal antibody specifically detects SARS-CoV-2 nsp3 in SARS-CoV-2 samples. It is validated for Western BlotSpecifications
| SARS-CoV-2 nsp3 | |
| Polyclonal | |
| Purified | |
| RUO | |
| SARS-CoV-2 | |
| Non-structural protein 3, Papain-like protease, Papain-like proteinase, PL2-PRO, PL-PRO, SARS-CoV-2NSP3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-199 of coronavirus NSP3 (YP_009725299.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Infections (Virus Bacteria and Parasites) | |
| PBS, 50% glycerol, pH7.3 | |
| 43740578 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title