missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 nsp15 Rabbit anti-SARS-CoV-2, Polyclonal, Novus Biologicals™

Product Code. 18308396 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18308396 20 μg 20µL
18372724 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18308396 Supplier Novus Biologicals Supplier No. NBP31599220UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 nsp15 Polyclonal antibody specifically detects SARS-CoV-2 nsp15 in SARS-CoV-2 samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 nsp15
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias endoRNAse, Non-structural protein 15
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 6600-6700 of coronavirus NSP15 (YP_009724389.1).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 43740578
Target Species SARS-CoV-2
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.