missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 nsp14 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18389211 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389211 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18389211 Supplier Novus Biologicals Supplier No. NBP307054AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 nsp14 Polyclonal antibody specifically detects SARS-CoV-2 nsp14 in SARS-CoV-2 samples. It is validated for ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 nsp14
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ExoN, Guanine-N7 methyltransferase, Non-structural protein 14, nsp14, Proofreading exoribonuclease
Host Species Rabbit
Immunogen This antibody was raised against a peptide corresponding to 14 amino acids near the center of SARS-CoV-2 (COVID-19) NSP14 (ExoN) protein. The immunogen is located between 30-80 amino acids of SARS-CoV-2 (COVID-19) NSP14 (ExoN).
Purification Method Peptide Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 43740578
Target Species SARS-CoV-2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.