missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 NSP10 Rabbit anti-SARS-CoV-2, Polyclonal, Novus Biologicals™

Product Code. 18321903 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321903 100 μg 100µL
18357223 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18321903 Supplier Bio-Techne Supplier No. NBP316002100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 NSP10 Polyclonal antibody specifically detects SARS-CoV-2 NSP10 in SARS-CoV-2 samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 NSP10
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias GFL, Growth factor-like peptide, Non-structural protein 10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-139 of coronavirus NSP10 (YP_009725306.1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 43740578
Target Species SARS-CoV-2
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.