missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ SARS-CoV-2 N protein ELISA Kit
Shop All Proteintech Products
missing translation for 'orderingAttributeHoverText'
Assay Sensitivity:
38 pg/mL
missing translation for 'unitSize'
96 tests
Description
- KE30007 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA)
- The SARS-CoV-2 N protein ELISA kit is to be used to detect and quantify protein levels of endogenous SARS-CoV-2 N protein
- The assay recognizes human SARS-CoV-2 N protein. An antibody specific for SARS-CoV-2 N protein has been pre-coated onto the microwells
- The SARS-CoV-2 N protein in samples is captured by the coated antibody after incubation
- Following extensive washing, another antibody specific for SARS-CoV-2 N protein is added to detect the captured SARS-CoV-2 N protein
- For signal development, horseradish peroxidase (HRP)-conjugated antibody is added, followed by Tetramethyl-benzidine (TMB) reagent
- Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm.
Spécification
Spécification
| Accession Number | NC_045512 |
| Assay Range | 375 to 6000 pg/mL |
| Assay Sensitivity | 38 pg/mL |
| Certifications/Compliance | ISO 13485 |
| Detection Method | Sandwich ELISA, Double Antibody |
| Immunoassay Kit Format | Coated ELISA Kit |
| Sample Type | Plasma, Serum |
| Substrate | DAB |
| Immunoassay Kit Grade | Solid |
| Interassay CV | <10% |
| Afficher plus |
Sécurité et traitement
Identifiant produit
- 96TEST SARS-CoV-2 N protein ELISA Kit
- Warning
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P234-Keep only in original container.
- P264-Wash skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P330-Rinse mouth.
- P362-Take off contaminated clothing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu