missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 Membrane Protein Antibody, DyLight 550, Novus Biologicals™

Product Code. 18334573
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18334573 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18334573 Supplier Bio-Techne Supplier No. NBP307059R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 Membrane Protein Polyclonal antibody specifically detects SARS-CoV-2 Membrane Protein in SARS-CoV-2 samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 Membrane Protein
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 2019-nCoV M protein, 2019-nCoV Membrane Protein, COVID-19 E1 Glycoprotein, COVID-19 Matrix glycoprotein, COVID-19 Membrane, COVID-19 Membrane glycoprotein, E1 glycoprotein, Human coronavirus Membrane Protein, Matrix glycoprotein, Membrane glycoprotein, Membrane Protein, SARS-CoV-2, SARS-CoV-2 E1 Glycoprotein, SARS-CoV-2 M Protein, SARS-CoV-2 Matrix glycoprotein, SARS-CoV-2 Membrane glycoprotein, SARSCoV2 Membrane Protein, Severe Acute Respiratory Syndrome Coronavirus 2
Host Species Rabbit
Immunogen This antibody was raised against a peptide corresponding to 15 amino acids near the carboxyl terminus of SARS-CoV-2 (COVID-19) Membrane protein. The immunogen is located within the last 50 amino acids of SARS-CoV-2 (COVID-19) Membrane protein.
Purification Method Peptide Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 43740571
Target Species SARS-CoV-2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.