missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 Membrane Protein Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18399432 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399432 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18399432 Supplier Novus Biologicals Supplier No. NBP307058AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 Membrane Protein Polyclonal antibody specifically detects SARS-CoV-2 Membrane Protein in SARS-CoV-2 samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 Membrane Protein
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 2019-nCoV M protein, 2019-nCoV Membrane Protein, COVID-19 E1 Glycoprotein, COVID-19 Matrix glycoprotein, COVID-19 Membrane, COVID-19 Membrane glycoprotein, E1 glycoprotein, Human coronavirus Membrane Protein, Matrix glycoprotein, Membrane glycoprotein, Membrane Protein, SARS-CoV-2, SARS-CoV-2 E1 Glycoprotein, SARS-CoV-2 M Protein, SARS-CoV-2 Matrix glycoprotein, SARS-CoV-2 Membrane glycoprotein, SARSCoV2 Membrane Protein, Severe Acute Respiratory Syndrome Coronavirus 2
Host Species Rabbit
Immunogen This antibody was raised against a peptide corresponding to 13 amino acids near the center of SARS-CoV-2 (COVID-19) Membrane protein. The immunogen is located between 130-180 amino acids of the SARS-CoV-2 (COVID-19) Membrane protein.
Purification Method Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 43740571
Target Species SARS-CoV-2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.