missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 Envelope Protein Rabbit Polyclonal Antibody, Proteintech

Product Code. 30235674 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
Unit Size:
150µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30235674 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30235674 Supplier Proteintech Supplier No. 289041AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The SARS-CoV-2 Envelope Protein antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of virus SARS-CoV-2 Envelope Protein. This antibody recognizes virus antigen. The SARS-CoV-2 Envelope Protein antibody has been validated for the following applications: ELISA analysis.

Specifications

Antigen SARS-CoV-2 Envelope Protein
Applications ELISA
Classification Polyclonal
Concentration 575 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Immunogen Recombinant Protein
Purification Method Antigen affinity purification
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740570
Target Species Virus
Content And Storage Store at -20°C. Stable for one year after shipment.
Product Type Polyclonal Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.