missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 Envelope Antibody, HRP, Novus Biologicals™

Product Code. 18345552
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18345552 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18345552 Supplier Bio-Techne Supplier No. NBP307060H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 Envelope Polyclonal antibody specifically detects SARS-CoV-2 Envelope in SARS-CoV-2 samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 Envelope
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate HRP
Formulation PBS without Preservative
Gene Alias 2019-nCoV Envelope protein, COVID-19 Envelope protein, E Protein, Envelope, Human Coronavirus Envelope protein, SARSCoV2, SARS-CoV-2 E protein, SARSCoV2 Envelope protein, SARS-CoV-2 Envelope protein, Severe acute respiratory syndrome 2 Envelope Protein
Host Species Rabbit
Immunogen This antibody was raised against a peptide corresponding to 14 amino acids near the carboxyl terminus of SARS-CoV-2 (COVID-19) Envelope protein. The immunogen is located within the last 50 amino acids of SARS-CoV-2 (COVID-19) Envelope protein.
Purification Method Peptide Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740570
Target Species SARS-CoV-2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.