Learn More
Invitrogen™ SARS-CoV-2 Delta Spike Protein RBD Chimeric Recombinant Human Monoclonal Antibody (D006)
GREENER_CHOICE
Human Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA560606
 View more versions of this product
Product Code. 30534332
  • £473.00 / 100µg
Description
Specifically recognize the SARS-CoV-2 Delta Spike RBD and ECD. No cross-reactivity with SARS-CoV-2 wild type, Alpha, Beta, Omicron BA.2.75 variant.. Has cross-reactivity in ELISA with: SARS-CoV-2 Delta (B.1.617.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Delta (B.1.617.2) Spike RBD Protein; No cross-reactivity in ELISA with: SARS-CoV-2 Omicron (XBB.1.16) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB.1.5) Spike RBD Protein; SARS-CoV-2 Omicron (XBB.1.5) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB.1) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (XBB) Spike RBD Protein; SARS-CoV-2 Omicron (BQ.1.1) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BQ.1.1)Spike RBD Protein; SARS-CoV-2 Omicron (BF.7) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4.6/BF.7) Spike RBD Protein; SARS-CoV-2 Omicron (BA.4.6) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.2.75) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.2.75.2) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.75.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4/BA.5/BA.5.2) Spike S1+S2 trimer Protein; SARS-CoV-2 Omicron (BA.4/BA.5/BA.5.2) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.75) Spike RBD Protein; SARS-CoV-2 Omicron (BA.2.3.20) Spike RBD Protein; SARS-CoV-2 Omicron (B.1.1.529) S1+S2 trimer Protein; SARS-CoV-2 Omicron (B.1.1.529) Spike RBD Protein; SARS-CoV-2 Alpha (B.1.1.7) Spike S1+S2 trimer Protein; SARS-CoV-2 Alpha (B.1.1.7) Spike RBD Protein; SARS-CoV-2 Beta (B.1.351) Spike S1+S2 trimer Protein; S...
The SARS-CoV-2 Delta Spike Protein RBD refers to the receptor-binding domain (RBD) of the spike (S) protein in the Delta variant of the SARS-CoV-2 virus, which is responsible for COVID-19. The RBD is a critical region within the S1 subunit of the spike protein that binds to the angiotensin-converting enzyme 2 (ACE2) receptor on human cells, facilitating viral entry. The Delta variant, first identified in India, possesses several mutations in the RBD that can enhance its binding affinity to the ACE2 receptor, potentially increasing the virus's transmissibility and partially evading immune responses. These mutations have significant implications for the effectiveness of vaccines and therapeutic antibodies, making it essential to monitor and adapt public health strategies accordingly.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SARS-CoV-2 Delta Spike Protein RBD Chimeric
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
Recombinant SARS-CoV-2 Delta Spike ECD Protein.
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Neutralization
D006
Unconjugated
Human
Protein A
RUO
Virus
Antibody
IgG1 κ