missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems SARS-CoV-2 (COVID-19) Spike S1 protein-coupled magnetic beads

Product Code. 17136481
Click to view available options
Quantity:
2 mg
10 mg
Unit Size:
10mg
2mg

Product Code. 17136481

Brand: ACROBiosystems MBSK00110mg Beads

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

For capturing the Anti- SARS-CoV-2 antibody or ACE2 protein from cell or serum sample

The pre-coupled magnetic beads coupled with biotinylated SARS-CoV-2 Spike S1 protein to streptavidin conjugated magnetic beads, which can capture the Anti- SARS-CoV-2 antibody or ACE2 protein from cell or serum sample. The beads are in uniform size, narrow size distribution with large surface area and unique surface coating, which can help you get the best performance and highly reproducible results. This very first SARS-CoV-2 Spike protein S1-coupled magnetic beads will bring great convenience with minimum non-specific binding and developed protocols. This ready to use products could greatly save your time and hassle.

Specifications

Kit Contents The Antigen pre-Coupled magnetic beads are Coupled with Biotinylated protein onto streptavidin (SA) magnetic beads. Because Streptavidin (SA) has an extraordinarily high affinity for biotin with a dissociation constant (Kd) on the order of 10-14 mol/L, th
Content And Storage -20°C, Avoid light
Shipping Condition RT
Quantity 10 mg
Type Magnetic Affinity Bead
Species SARS-CoV-2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.