missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 3CL Protease Antibody, Novus Biologicals™
Description
SARS-CoV-2 3CL Protease Polyclonal antibody specifically detects SARS-CoV-2 3CL Protease in SARS-CoV-2 samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | SARS-CoV-2 3CL Protease |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 20% Glycerol (pH7) |
| Gene Alias | 3CL Proteinase, 3C-like main protease, 3CL-Mpro, COVID-19, M Proteinase |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the center region of SARS-CoV-2 (COVID-19) 3CLpro (nsp5). The exact sequence is proprietary. |
| Purification Method | Affinity purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?