missing translation for 'onlineSavingsMsg'
Learn More

SARS-CoV-2 3CL Protease Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18341742 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341742 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341742 Supplier Novus Biologicals Supplier No. NBP307061AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SARS-CoV-2 3CL Protease Polyclonal antibody specifically detects SARS-CoV-2 3CL Protease in SARS-CoV-2 samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 3CL Protease
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 3CL Proteinase, 3C-like main protease, 3CL-Mpro, COVID-19, M Proteinase
Host Species Rabbit
Immunogen This antibody was raised against a peptide corresponding to 15 amino acids in the center of SARS-CoV-2 (COVID-19) 3CL-PRO (NSP5) protein. The immunogen is located within 80-130 amino acids of the SARS-CoV-2 (COVID-19) 3CL-PRO (NSP5) protein.
Purification Method Peptide Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740578
Target Species SARS-CoV-2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.