missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS Coronavirus Spike Protein Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120761
This item is not returnable.
View return policy
Description
Positive test controls include: 293T. Immunogen sequence: DVNCTDVSTA IHADQLTPAW RIYSTGNNVF QTQAGCLIGA EHVDTSYECD IPIGAGICAS YHTVSLLRST SQKSIVAYTM SLGADSSIAY SNNTIAIPTN F.
It has recently been shown that SARS (severe acute respiratory syndrome) is caused by a human coronavirus. Human coronaviruses are the major cause of upper respiratory tract illness, such as the common cold, in humans. Coronaviruses are positive-stranded RNA viruses, featuring the largest viral RNA genomes known to date (27-31 kb). The first step in coronavirus infection is binding of the viral spike protein, a 139-kDa protein, to certain receptors on host cells. The spike protein is the main surface antigen of the coronavirus. The glycosylated spike protein (as well as the nucleocapsid protein) can be detected in infected cell culture supernatants with antisera from SARS patients.
Specifications
| SARS Coronavirus Spike Protein | |
| Polyclonal | |
| Unconjugated | |
| Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2 | |
| A synthetic peptide corresponding to a sequence within amino acids 600-700 of coronavirus Spike (NP_8288511). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.04 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction