missing translation for 'onlineSavingsMsg'
Learn More

Sarolaner, MedChemExpress

Product Code. 16473187
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473187 50 mg 50mg
16467156 100 mg 100mg
16429697 5 mg 5mg
16427168 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16473187 Supplier Medchem Express Supplier No. HY16730

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sarolaner (PF-6450567) is an orally active and broad-spectrum ectoparasiticide. Sarolaner is an isoxazoline compound which shows efficacy against fleas and ticks on dogs, with a LC80 value of 0.3 μg/mL against C. felis and a LC100 value of 0.003 μg/mL against O. turicata.

Sarolaner (PF-6450567) is an orally active and broad-spectrum ectoparasiticide. Sarolaner is an isoxazoline compound which shows efficacy against fleas and ticks on dogs, with a LC80 value of 0.3 μg/mL against C. felis and a LC100 value of 0.003 μg/mL against O. turicata.

Specifications

Chemical Name or Material Sarolaner
Physical Form Solid
CAS 1398609-39-6
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C23H18Cl2F4N2O5S
For Use With (Application) COVID-19-immunoregulation
Synonym PF-6450567
Solubility Information DMSO : 300 mg/mL (516.03 mM; Need ultrasonic)
SMILES O=S(CC(N1CC2(OCC3=C2C=CC(C4=NO[C@](C(F)(F)F)(C5=CC(Cl)=C(F)C(Cl)=C5)C4)=C3)C1)=O)(C)=O
Molecular Weight (g/mol) 581.36
Formula Weight 581.36
Percent Purity 98.08%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.