missing translation for 'onlineSavingsMsg'
Learn More

Sarolaner, MedChemExpress

Product Code. 16429697
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16429697

Brand: Medchem Express HY16730

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sarolaner (PF-6450567) is an orally active and broad-spectrum ectoparasiticide. Sarolaner is an isoxazoline compound which shows efficacy against fleas and ticks on dogs, with a LC80 value of 0.3 μg/mL against C. felis and a LC100 value of 0.003 μg/mL against O. turicata.

Sarolaner (PF-6450567) is an orally active and broad-spectrum ectoparasiticide. Sarolaner is an isoxazoline compound which shows efficacy against fleas and ticks on dogs, with a LC80 value of 0.3 μg/mL against C. felis and a LC100 value of 0.003 μg/mL against O. turicata.

Specifications

Chemical Name or Material Sarolaner
Physical Form Solid
CAS 1398609-39-6
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C23H18Cl2F4N2O5S
For Use With (Application) COVID-19-immunoregulation
Synonym PF-6450567
Solubility Information DMSO : 300 mg/mL (516.03 mM; Need ultrasonic)
SMILES O=S(CC(N1CC2(OCC3=C2C=CC(C4=NO[C@](C(F)(F)F)(C5=CC(Cl)=C(F)C(Cl)=C5)C4)=C3)C1)=O)(C)=O
Molecular Weight (g/mol) 581.36
Formula Weight 581.36
Percent Purity 98.08%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.