missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saroglitazar Magnesium, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
Description
Saroglitazar magnesium is a novel peroxisome proliferator-activated receptor (PPAR) agonist with predominant PPARα and moderate PPARγ activity with EC50 values of 0.65 pM and 3 nM in HepG2 cells, respectively.
Specifications
Specifications
| Chemical Name or Material | Saroglitazar Magnesium |
| Physical Form | Solid |
| CAS | 1639792-20-3 |
| Color | Light Yellow |
| Quantity | 1 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C50H56MgN2O8S2 |
| For Use With (Application) | Metabolism-sugar/lipid metabolism |
| Solubility Information | DMSO : 125 mg/mL (138.67 mM; Need ultrasonic) |
| SMILES | CCO([Mg+2]1(O(CC)[C@H](C([O-]1)=O)CC2=CC=C(OCCN3C(C)=CC=C3C4=CC=C(SC)C=C4)C=C2)[O-]C5=O)[C@H]5CC6=CC=C(OCCN7C(C)=CC=C7C8=CC=C(SC)C=C8)C=C6 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction