missing translation for 'onlineSavingsMsg'
Learn More

Saroglitazar Magnesium, MedChemExpress

Product Code. 16497968
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16497968

Brand: Medchem Express HY19937A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Saroglitazar magnesium is a novel peroxisome proliferator-activated receptor (PPAR) agonist with predominant PPARα and moderate PPARγ activity with EC50 values of 0.65 pM and 3 nM in HepG2 cells, respectively.

Saroglitazar magnesium is a novel peroxisome proliferator-activated receptor (PPAR) agonist with predominant PPARα and moderate PPARγ activity with EC50 values of 0.65 pM and 3 nM in HepG2 cells, respectively.

Specifications

Chemical Name or Material Saroglitazar Magnesium
Physical Form Solid
CAS 1639792-20-3
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C50H56MgN2O8S2
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information DMSO : 125 mg/mL (138.67 mM; Need ultrasonic)
SMILES CCO([Mg+2]1(O(CC)[C@H](C([O-]1)=O)CC2=CC=C(OCCN3C(C)=CC=C3C4=CC=C(SC)C=C4)C=C2)[O-]C5=O)[C@H]5CC6=CC=C(OCCN7C(C)=CC=C7C8=CC=C(SC)C=C8)C=C6
Molecular Weight (g/mol) 901.42
Formula Weight 901.42
Percent Purity 98.03%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.