missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARNP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521783
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (95%), Rat (95%), Chicken (80%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein that is upregulated in response to various cytokines. The encoded protein may play a role in cell cycle progression. A translocation between this gene and the myeloid/lymphoid leukemia gene, resulting in expression of a chimeric protein, has been associated with acute myelomonocytic leukemia. Pseudogenes exist on chromosomes 7 and 8. Alternatively spliced transcript variants have been described.
Specifications
| SARNP | |
| Polyclonal | |
| Unconjugated | |
| SARNP | |
| 1110005A23Rik; CIP29; cytokine induced protein 29 kDa; cytokine-induced protein of 29 kDa; HCC1; hepatocellular carcinoma 1; HSPC316; Nuclear protein Hcc-1; proliferation associated cytokine-inducible protein CIP29; proliferation-associated cytokine-inducible protein CIP29; RGD1305692; SAP domain containing ribonucleoprotein; SAP domain-containing ribonucleoprotein; Sarnp; THO1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84324 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.33 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P82979 | |
| SARNP | |
| Recombinant protein encompassing a sequence within the center region of human CIP29. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction