missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SARM1 Polyclonal antibody specifically detects SARM1 in Human, Drosophila samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | SARM1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 20% Glycerol (pH7) |
| Gene Alias | KIAA0524SAM domain-containing protein 2, SAMD2sterile alpha and HEAT/Armadillo motif protein, ortholog of Drosophila, SARMFLJ36296, Sterile alpha and Armadillo repeat protein, sterile alpha and TIR motif containing 1, sterile alpha and TIR motif-containing protein 1, Sterile alpha motif domain-containing protein 2, Tir-1 homolog |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human SARM1. The exact sequence is proprietary. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?