missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARM1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£141.00 - £398.00
Specifications
| Antigen | SARM1 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18638218
|
Novus Biologicals
NBP2-41180-0.025mg |
0.025 mg |
£141.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18153859
|
Novus Biologicals
NBP2-41180 |
0.1 mg |
£398.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SARM1 Polyclonal specifically detects SARM1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| SARM1 | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 23098 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human SARM. The immunogen is located within amino acids 30 - 80 of SARM. Amino Acid Squence: AGGRGPREVSPGAGTE | |
| Primary | |
| At least three alternatively spliced transcript variants encoding distinct isoforms have been observed. SARM antibody recognize the longest isoform. |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| KIAA0524SAM domain-containing protein 2, SAMD2sterile alpha and HEAT/Armadillo motif protein, ortholog of Drosophila, SARMFLJ36296, Sterile alpha and Armadillo repeat protein, sterile alpha and TIR motif containing 1, sterile alpha and TIR motif-containing protein 1, Sterile alpha motif domain-containing protein 2, Tir-1 homolog | |
| SARM1 | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title