missing translation for 'onlineSavingsMsg'
Learn More

SARM1 Antibody [Biotin], Novus Biologicals™

Product Code. 30731042 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731042 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731042 Supplier Novus Biologicals Supplier No. AF7037B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

SARM1 Polyclonal antibody specifically detects SARM1 in Human samples. It is validated for Immunoprecipitation,Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARM1
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias KIAA0524SAM domain-containing protein 2, SAMD2sterile alpha and HEAT/Armadillo motif protein, ortholog of Drosophila, SARMFLJ36296, Sterile alpha and Armadillo repeat protein, sterile alpha and TIR motif containing 1, sterile alpha and TIR motif-containing protein 1, Sterile alpha motif domain-containing protein 2, Tir-1 homolog
Host Species Sheep
Immunogen E. coli-derived recombinant human SARM1, Lys556-Thr724, Accession # Q6SZW1
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 23098
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.