missing translation for 'onlineSavingsMsg'
Learn More

Saridegib, MedChemExpress

Product Code. p-200096377
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16487088

Brand: Medchem Express HY16587

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Saridegib is a potent and specific inhibitor of Smoothened (Smo), a key signaling transmembrane protein in the Hedgehog (Hh) pathway.

Saridegib is a potent and specific inhibitor of Smoothened (Smo), a key signaling transmembrane protein in the Hedgehog (Hh) pathway.

Specifications

Chemical Name or Material Saridegib
CAS 1037210-93-7
Color White
Purity Grade Notes Research
Molecular Formula C29H48N2O3S
Quantity 10 mg
Synonym IPI-926 Patidegib
SMILES CC1=C(C[C@@]2([H])[C@@]3([H])CC[C@@]4([H])[C@]2(C)CC[C@@H](NS(C)(=O)=O)C4)[C@@]3([H])CC[C@@]([C@@H]5C)(C1)O[C@@]6([H])[C@@]5([H])NC[C@@H](C)C6
Molecular Weight (g/mol) 504.77
Formula Weight 504.77
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.