missing translation for 'onlineSavingsMsg'
Learn More

Saridegib, MedChemExpress

Product Code. 16429637
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16429637 5 mg 5mg
16487088 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16429637 Supplier Medchem Express Supplier No. HY16587
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Saridegib is a potent and specific inhibitor of Smoothened (Smo), a key signaling transmembrane protein in the Hedgehog (Hh) pathway.

Saridegib is a potent and specific inhibitor of Smoothened (Smo), a key signaling transmembrane protein in the Hedgehog (Hh) pathway.

Specifications

Chemical Name or Material Saridegib
CAS 1037210-93-7
Color White
Purity Grade Notes Research
Molecular Formula C29H48N2O3S
Quantity 5 mg
Synonym IPI-926 Patidegib
SMILES CC1=C(C[C@@]2([H])[C@@]3([H])CC[C@@]4([H])[C@]2(C)CC[C@@H](NS(C)(=O)=O)C4)[C@@]3([H])CC[C@@]([C@@H]5C)(C1)O[C@@]6([H])[C@@]5([H])NC[C@@H](C)C6
Molecular Weight (g/mol) 504.77
Formula Weight 504.77
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.